Assay Details
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015100465.2 | XP_014955951.2 | 4 - 5 | 685 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
