Assay Details
Gene Symbol: LOC114112997 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964563.1 | XP_027820364.1 | 2 - 3 | 180 | 101 |
Gene Symbol: LOC101121659 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964562.1 | XP_027820363.1 | 2 - 3 | 161 | 101 |

More Information
Set Membership: |
![]() ![]() |
