Assay Details
| Gene Symbol: HINFP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979292.1 | XP_027835093.1 | 7 - 8 | 1015 | 63 | |
| XM_027979293.1 | XP_027835094.1 | 7 - 8 | 1183 | 63 | ||
More Information
Set Membership: |
|

HINFP
Genomic Map