Assay Details
Gene Symbol: LOC101106719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957647.1 | XP_027813448.1 | 10 - 11 | 1653 | 88 | |
XM_027957648.1 | XP_027813449.1 | 11 - 12 | 1542 | 88 | ||
XM_027957649.1 | XP_027813450.1 | 10 - 11 | 1653 | 88 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
