Assay Details
Gene Symbol: LOC101106719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957647.1 | XP_027813448.1 | 26 - 27 | 4709 | 74 | |
XM_027957648.1 | XP_027813449.1 | 27 - 28 | 4598 | 74 | ||
XM_027957649.1 | XP_027813450.1 | 26 - 27 | 4709 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
