Assay Details
Gene Symbol: LOC101106719 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957647.1 | XP_027813448.1 | 29 - 30 | 5235 | 65 | |
XM_027957648.1 | XP_027813449.1 | 30 - 31 | 5124 | 65 | ||
XM_027957649.1 | XP_027813450.1 | 29 - 30 | 5229 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
