Assay Details
| Gene Symbol: CEMIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004310.4 | XP_004004359.2 | 6 - 7 | 1576 | 53 | |
| XM_027964507.1 | XP_027820308.1 | 6 - 7 | 1576 | 53 | ||
More Information
Set Membership: |
|

CEMIP2
Genomic Map