Assay Details
| Gene Symbol: DHRS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975317.1 | XP_027831118.1 | 6 - 7 | 1000 | 73 | |
| XM_027975318.1 | XP_027831119.1 | 5 - 6 | 874 | 73 | ||
More Information
Set Membership: |
|

DHRS11
Genomic Map