Assay Details
| Gene Symbol: CNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974523.1 | XP_027830324.1 | 3 - 4 | 912 | 94 | |
| XM_027974524.1 | XP_027830325.1 | 3 - 4 | 839 | 94 | ||
| GenBank mRNA | JN684760.1 | - | 2 - 3 | 808 | 94 | |
More Information
Set Membership: |
|

CNP
Genomic Map