Assay Details
Gene Symbol: LOC101116577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012182163.3 | XP_012037553.1 | 4 - 5 | 1465 | 63 | |
XM_027972087.1 | XP_027827888.1 | 4 - 5 | 1465 | 63 | ||
XM_027972088.1 | XP_027827889.1 | 13 - 14 | 3836 | 63 | ||
XM_027972089.1 | XP_027827890.1 | 4 - 5 | 1465 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
