Assay Details
Gene Symbol: LOC101116577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012182163.3 | XP_012037553.1 | 5 - 6 | 1772 | 81 | |
XM_027972087.1 | XP_027827888.1 | 5 - 6 | 1772 | 81 | ||
XM_027972088.1 | XP_027827889.1 | 14 - 15 | 4143 | 81 | ||
XM_027972089.1 | XP_027827890.1 | 5 - 6 | 1772 | 81 |
