Assay Details
| Gene Symbol: RNF114 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014631.4 | XP_004014680.1 | 5 - 6 | 646 | 67 | |
| XM_027977313.1 | XP_027833114.1 | 6 - 7 | 792 | 67 | ||
More Information
Set Membership: |
|

RNF114
Genomic Map