Assay Details
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018467.4 | XP_004018516.2 | 5 - 6 | 1840 | 68 | |
XM_027957644.1 | XP_027813445.1 | 5 - 6 | 1099 | 68 | ||
XM_027957645.1 | XP_027813446.1 | 6 - 7 | 1217 | 68 | ||
XM_027957646.1 | XP_027813447.1 | 5 - 6 | 1195 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
