Assay Details
Gene Symbol: SKIL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958415.1 | XP_027814216.1 | 1 - 2 | 2792 | 83 | |
XM_027958421.1 | XP_027814222.1 | 1 - 2 | 2792 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
