Assay Details
Gene Symbol: LOC101115706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969603.1 | XP_027825404.1 | 25 - 26 | 3284 | 66 | |
XM_027969604.1 | XP_027825405.1 | 24 - 25 | 3058 | 66 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
