Assay Details
| Gene Symbol: DMAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015271.3 | XP_004015320.2 | 2 - 3 | 260 | 78 | |
| XM_027978348.1 | XP_027834149.1 | 2 - 3 | 260 | 78 | ||
More Information
Set Membership: |
|

DMAC2
Genomic Map