Assay Details
Gene Symbol: LOC114112072 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963720.1 | XP_027819521.1 | 1 - 2 | 48 | 62 |
Gene Symbol: ANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012179446.3 | XP_012034836.2 | 20 - 21 | 3638 | 62 | |
XM_027970795.1 | XP_027826596.1 | 20 - 21 | 2575 | 62 | ||
XM_027970796.1 | XP_027826597.1 | 19 - 20 | 2551 | 62 | ||
XM_027970797.1 | XP_027826598.1 | 20 - 21 | 2574 | 62 | ||
XM_027970798.1 | XP_027826599.1 | 19 - 20 | 2550 | 62 | ||
XM_027970799.1 | XP_027826600.1 | 19 - 20 | 2476 | 62 | ||
XM_027970800.1 | XP_027826601.1 | 19 - 20 | 2475 | 62 | ||
XM_027970801.1 | XP_027826602.1 | 18 - 19 | 2451 | 62 | ||
XM_027970802.1 | XP_027826603.1 | 17 - 18 | 2352 | 62 | ||
XM_027970803.1 | XP_027826604.1 | 20 - 21 | 2576 | 62 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
