Assay Details
| Gene Symbol: MAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011594.3 | XP_004011643.1 | 2 - 3 | 494 | 60 | |
| XM_012183441.2 | XP_012038831.1 | 2 - 3 | 616 | 60 | ||
| XM_027973432.1 | XP_027829233.1 | 1 - 2 | 582 | 60 | ||
| XM_027973433.1 | XP_027829234.1 | 2 - 3 | 486 | 60 | ||
More Information
Set Membership: |
|

MAF1
Genomic Map