Assay Details
| Gene Symbol: ACSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012763.4 | XP_004012812.2 | 12 - 13 | 1867 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACSF2| Gene Symbol: ACSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012763.4 | XP_004012812.2 | 12 - 13 | 1867 | 60 | |
Set Membership: |
|