Assay Details
Gene Symbol: LOC101117577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965980.1 | XP_027821781.1 | 14 - 15 | 1704 | 90 |
Gene Symbol: LOC114113811 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965979.1 | XP_027821780.1 | 3 - 4 | 372 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
