Assay Details
Gene Symbol: LOC101117577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965980.1 | XP_027821781.1 | 17 - 18 | 2015 | 70 |
Gene Symbol: LOC114113811 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965979.1 | XP_027821780.1 | 6 - 7 | 683 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
