Assay Details
Gene Symbol: CASP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008465.4 | XP_004008514.1 | 6 - 7 | 842 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
