Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006690.4 | XP_004006739.2 | 19 - 20 | 1950 | 103 | |
XM_027967682.1 | XP_027823483.1 | 12 - 13 | 1379 | 103 |

More Information
Set Membership: |
![]() ![]() |
