Assay Details
Gene Symbol: NSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971303.1 | XP_027827104.1 | 9 - 10 | 2565 | 56 | |
XM_027971304.1 | XP_027827105.1 | 8 - 9 | 2179 | 56 | ||
XM_027971305.1 | XP_027827106.1 | 9 - 10 | 1845 | 56 | ||
XM_027971306.1 | XP_027827107.1 | 9 - 10 | 2582 | 56 | ||
XM_027971307.1 | XP_027827108.1 | 8 - 9 | 2159 | 56 | ||
XM_027971308.1 | XP_027827109.1 | 8 - 9 | 1833 | 56 | ||
XM_027971309.1 | XP_027827110.1 | 9 - 10 | 2575 | 56 | ||
XM_027971310.1 | XP_027827111.1 | 9 - 10 | 2579 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
