Assay Details
Gene Symbol: MTUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012184654.3 | XP_012040044.2 | 8 - 9 | 1182 | 64 | |
XM_027973632.1 | XP_027829433.1 | 16 - 17 | 5011 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
