Assay Details
| Gene Symbol: LOC101104528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956323.1 | XP_027812124.1 | 42 - 43 | 7325 | 98 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101104528| Gene Symbol: LOC101104528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956323.1 | XP_027812124.1 | 42 - 43 | 7325 | 98 | |