Assay Details
| Gene Symbol: WSB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017380.4 | XP_004017429.1 | 6 - 7 | 962 | 59 | |
| XM_015101647.2 | XP_014957133.1 | 7 - 8 | 1112 | 59 | ||
| GenBank mRNA | HM589038.1 | - | 2 - 3 | 279 | 59 | |
More Information
Set Membership: |
|

WSB2
Genomic Map