Assay Details
Gene Symbol: TIPRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002745.4 | XP_004002794.1 | 2 - 3 | 419 | 106 | |
XM_012185836.3 | XP_012041226.1 | 2 - 3 | 523 | 106 | ||
XM_027977940.1 | XP_027833741.1 | 2 - 3 | 439 | 106 | ||
XM_027977950.1 | XP_027833751.1 | 2 - 3 | 445 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
