Assay Details
Gene Symbol: LOC114112156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963732.1 | XP_027819533.1 | 2 - 3 | 340 | 85 |
Gene Symbol: DHX57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966915.1 | XP_027822716.1 | 4 - 5 | 632 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
