Assay Details
Gene Symbol: WASHC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006690.4 | XP_004006739.2 | 24 - 25 | 2578 | 143 | |
XM_027967682.1 | XP_027823483.1 | 17 - 18 | 2007 | 143 |

More Information
Set Membership: |
![]() ![]() |
