Assay Details
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018994.4 | XP_004019043.1 | 2 - 3 | 223 | 57 | |
XM_012100976.3 | XP_011956366.1 | 2 - 3 | 195 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
