Assay Details
| Gene Symbol: RPL38 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098980.2 | XP_014954466.1 | 3 - 4 | 189 | 97 | |
| XM_027974023.1 | XP_027829824.1 | 3 - 4 | 172 | 97 | ||
| XM_027974024.1 | XP_027829825.1 | 3 - 4 | 115 | 97 | ||
More Information
Set Membership: |
|

RPL38
Genomic Map