Assay Details
| Gene Symbol: NFE2L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012132935.3 | XP_011988325.2 | 3 - 4 | 551 | 68 | |
| XM_012132949.2 | XP_011988339.2 | 3 - 4 | 532 | 68 | ||
| XM_012132956.3 | XP_011988346.2 | 3 - 4 | 1060 | 68 | ||
| XM_015093345.2 | XP_014948831.2 | 3 - 4 | 592 | 68 | ||
More Information
Set Membership: |
|

NFE2L2
Genomic Map