Assay Details
| Gene Symbol: LOC101103602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004001935.4 | XP_004001984.1 | 14 - 15 | 1706 | 69 | |
| XM_012131207.3 | XP_011986597.1 | 13 - 14 | 1686 | 69 | ||
More Information
Set Membership: |
|

LOC101103602
Genomic Map