Assay Details
Gene Symbol: TSPAN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008009.4 | XP_004008058.1 | 2 - 3 | 224 | 65 | |
XM_027968759.1 | XP_027824560.1 | 3 - 4 | 736 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
