Assay Details
| Gene Symbol: C17H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980242.1 | XP_027836043.1 | 3 - 4 | 358 | 71 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C17H4orf45| Gene Symbol: C17H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027980242.1 | XP_027836043.1 | 3 - 4 | 358 | 71 | |