Assay Details
| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012105048.2 | XP_011960438.2 | 4 - 5 | 351 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF248| Gene Symbol: ZNF248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012105048.2 | XP_011960438.2 | 4 - 5 | 351 | 98 | |
Set Membership: |
|