Assay Details
| Gene Symbol: MAZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104163.2 | XP_014959649.2 | 4 - 5 | 1328 | 102 | |
| XM_027961913.1 | XP_027817714.1 | 4 - 5 | 1328 | 102 | ||
| XM_027961915.1 | XP_027817716.1 | 5 - 6 | 1279 | 102 | ||
More Information
Set Membership: |
|

MAZ
Genomic Map