Assay Details
| Gene Symbol: CIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957148.1 | XP_027812949.1 | 4 - 5 | 404 | 71 | |
| GenBank mRNA | FJ039530.1 | - | 3 - 4 | 316 | 71 | |
More Information
Set Membership: |
|

CIB2
Genomic Map