Assay Details
| Gene Symbol: PGAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974607.1 | XP_027830408.1 | 1 - 2 | 957 | 100 | |
| XM_027974608.1 | XP_027830409.1 | 1 - 2 | 966 | 100 | ||
| XM_027974609.1 | XP_027830410.1 | 1 - 2 | 991 | 100 | ||
More Information
Set Membership: |
|

PGAP3
Genomic Map