Assay Details
Gene Symbol: RIMS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015099894.2 | XP_014955380.2 | 5 - 6 | 596 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
