Assay Details
| Gene Symbol: LOC105610446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961957.1 | XP_027817758.1 | 11 - 12 | 615 | 115 | |
| Gene Symbol: LOC114110662 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961956.1 | XP_027817757.1 | 23 - 24 | 1098 | 115 | |
More Information
Set Membership: |
|

LOC105610446
Genomic Map