Assay Details
| Gene Symbol: LOC105610446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961957.1 | XP_027817758.1 | 19 - 20 | 933 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105610446| Gene Symbol: LOC105610446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961957.1 | XP_027817758.1 | 19 - 20 | 933 | 101 | |
Set Membership: |
|