Assay Details
Gene Symbol: LOC101117577 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965980.1 | XP_027821781.1 | 18 - 19 | 2124 | 85 |
Gene Symbol: LOC114113811 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965979.1 | XP_027821780.1 | 7 - 8 | 792 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
