Assay Details
Gene Symbol: LOC101119768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958604.1 | XP_027814405.1 | 4 - 5 | 525 | 77 |
Gene Symbol: LOC114109612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958668.1 | XP_027814469.1 | 4 - 5 | 525 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
