Assay Details
Gene Symbol: TIMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957804.1 | XP_027813605.1 | 3 - 4 | 424 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
