Assay Details
Gene Symbol: TMEM131L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956225.1 | XP_027812026.1 | 7 - 8 | 661 | 61 | |
XM_027956226.1 | XP_027812027.1 | 7 - 8 | 661 | 61 | ||
XM_027956227.1 | XP_027812028.1 | 7 - 8 | 661 | 61 | ||
XM_027956228.1 | XP_027812029.1 | 7 - 8 | 661 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
