Assay Details
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961242.1 | XP_027817043.1 | 7 - 8 | 1648 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961242.1 | XP_027817043.1 | 7 - 8 | 1648 | 91 |
Set Membership: |
![]() ![]() ![]() |