Assay Details
Gene Symbol: LOC106990117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958887.1 | XP_027814688.1 | 2 - 3 | 462 | 63 | |
XM_027958888.1 | XP_027814689.1 | 3 - 4 | 891 | 63 | ||
XM_027958890.1 | XP_027814691.1 | 2 - 3 | 457 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
