Assay Details
Gene Symbol: LOC106990117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958887.1 | XP_027814688.1 | 6 - 7 | 1224 | 90 | |
XM_027958888.1 | XP_027814689.1 | 7 - 8 | 1653 | 90 | ||
XM_027958889.1 | XP_027814690.1 | 6 - 7 | 1179 | 90 | ||
XM_027958890.1 | XP_027814691.1 | 6 - 7 | 1145 | 90 | ||
XM_027958891.1 | XP_027814692.1 | 5 - 6 | 947 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
